Learn More
Novus Biologicals™ Mouse IL-6 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192668
Catalog No. NBP192668
Add to cart
Description
Description
The Mouse IL-6 ELISA Kit (Colorimetric) measures mouse IL-6 in cell culture supernatant, serum, plasma (edta, citrate).
Specifications
Specifications
31.3 - 2,000pg/mL
3569
ELISA Kit
6.5pg/mL
Mouse (Mu)
IL-6
IL6
ELISA
Streptavidin-HRP, Mouse IL-6 Standard lyophilized, 4ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films, Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IL-6, Biotin-Conjugate anti-mouse IL-6 monoclonal antibody
96 Tests
Cell culture supernatant, serum, plasma (EDTA, citrate)
Storage of components varies. See protocol for specific instructions.
Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience
B cell stimulatory factor-2, B-cell differentiation factor, BSF-2, BSF2CTL differentiation factor, CDF, HGFHSFIFNB2Hybridoma growth factor, IFN-beta-2, IL-6B-cell stimulatory factor 2, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF