Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse IL-9 (aa 19-144), Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

Insect cells Recombinant Protein

$244.00 - $2060.00

Specifications

Recombinant Yes
Name Mouse IL-9 (aa 19-144)
For Use With (Application) Control, ELISA, Western Blot
Concentration 200 µg/mL
Conjugate Unlabeled
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87895 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87895
500 µg
Each for $2,060.00
Add to cart
 
PIRP87896 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87896
10 µg
Each for $244.00
Add to cart
 
PIRP87897 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87897
25 µg
Each for $481.50
Add to cart
 
Description

Description

Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Unlabeled
PBS with no preservative
Insect cells
-20°C
P15247
Il9
>95%
Mouse IL-9 (aa 19-144)
200 µg/mL
Liquid
cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; Il9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor p40
RUO
Mouse
16198
14 kD
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit