Learn More
Abnova™ Mouse Il19 (Q8CJ70, 152 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4473
Catalog No. 89-948-726
Add to cart
Specifications
Specifications
Q8CJ70
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
17.7kDa
Escherichia coli expression system
SDS-PAGE Stained with Coomassie Blue
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
Il19
Escherichia coli
None
Escherichia coli expression system
Lyophilized
329244
Il19 (Mouse) Recombinant Protein
≥95% by SDS-PAGE and HPLC
10 ug
RUO
Il19
Mouse IL-19 is fully biologically active when compared to standard. Significant STAT3 activation is observed with > 100ng/mL of recombinant mouse IL-19.
Yes