Learn More
Abnova™ Mouse Il31 (Q6EAL8) Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4632
Catalog No. 89-948-885
Add to cart
Description
Description
Sequence: MTCSLSFGAPISKEDLRTTIDLLKQESQDLYNNYSIKQASGMSADESIQLPCFSLDREALTNISVIIAHLEKVKVLSENTVDTSWVIRWLTNISCFNPLNLNISVPGNTDESYDCKVFVLTVLKQFSNCMAELQAKDNTTC
Specifications
Specifications
Q6EAL8
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
15.7kDa
Escherichia coli expression system
10 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1 EU/μg
Il31
The activity is determined by the dose dependent production of IL-8 by human PBMCs. The expected ED50 for this effect is 16-24pg/mL.
Yes
Escherichia coli expression system
Lyophilized
76399
Il31 (Mouse) Recombinant Protein
1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
MTCSLSFGAPISKEDLRTTIDLLKQESQDLYNNYSIKQASGMSADESIQLPCFSLDREALTNISVIIAHLEKVKVLSENTVDTSWVIRWLTNISCFNPLNLNISVPGNTDESYDCKVFVLTVLKQFSNCMAELQAKDNTTC
RUO
1700013B14Rik
Il31
Escherichia coli
None