Learn More
Abnova™ Mouse Il9 (P15247) Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4599
Catalog No. 89-948-967
Add to cart
Description
Description
Sequence: MQRCSTTWGIRDTNYLIENLKDDPPSKCSCSGNVTSCLCLSVPTDDCTTPCYREGLLQLTNATQKSRLLPVFHRVKRIVEVLKNITCPSFSCEKPCNQTMAGNTMSFLKSLLGTFQKTEMQRQKSRP
Specifications
Specifications
Escherichia coli expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
14.3kDa
Escherichia coli expression system
10 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1 EU/μg
Il9
The activity is determined by the dose-dependent proliferation of mouse Mo7e cells. The expected ED50 for this effect is 0.3-0.6ng/mL.
Yes
P15247
Lyophilized
16198
Il9 (Mouse) Recombinant Protein
1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
MQRCSTTWGIRDTNYLIENLKDDPPSKCSCSGNVTSCLCLSVPTDDCTTPCYREGLLQLTNATQKSRLLPVFHRVKRIVEVLKNITCPSFSCEKPCNQTMAGNTLSFLKSLLGTFQKTEMQRQKSRP
RUO
Il-9|P40
Il9
Escherichia coli
None