Learn More
Novus Biologicals™ Mouse iNOS ELISA Kit (Colorimetric)
This kit recognizes Mouse
Manufacturer:  Novus Biologicals™ NBP280256
Catalog No. NB013327
Add to cart
Description
Description
No significant cross-reactivity or interference between Mouse NOS2/iNOS and analogues was observed.
Specifications
Specifications
P29477, P29477
0.188 ng/mL
Serum, Plasma, Other Biological Fluids
ELISA
NOS2
1 Kit
Storage of components varies. See protocol for specific instructions.
0.313 to 20 ng/mL
ELISA Kit (Colorimetric)
iNOS
4843
Mouse
Adaptive Immunity, Apoptosis, Cancer, HIF Target Genes, Hypoxia, Inflammation, Lipid and Metabolism, Neuroscience, Neurotransmission
EC 1.14.13.39, Hepatocyte NOS, HEP-NOS, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS type II, NOS2A, Peptidyl-cysteine S-nitrosylase NOS2