Learn More
Novus Biologicals™ Mouse Insulin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP262853
Catalog No. NBP262853
Add to cart
Description
Description
The Mouse Insulin ELISA Kit (Colorimetric) measures mouse Insulin in serum, plasma and other biological fluids.
Specifications
Specifications
ELISA Kit (Colorimetric)
This kit recognizes Mouse INS in samples. No significant cross-reactivity or interference between Mouse INS and analogues was observed.
3630
Insulin
1 Kit
0.19 ng/mL
0.31 - 20 ng/mL
ELISA
Storage is content dependent.
Mouse
Biological Fluids, Plasma, Serum
INS