Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Mouse Kitl (182 a.a.) Partial Recombinant Protein with His tag expressed in Barley grain (Hordeum vulgare)

Used for Func, SDS-PAGE, WB-Re

Manufacturer:  Abnova™ P5931

Catalog No. 89-949-955

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Hordeum vulgare (barley) expression system
Lyophilized
17311
Mouse Kitl (182 a.a.) Partial Recombinant Protein with His tag expressed in Barley grain (Hordeum vulgare)
>95% by SDS-PAGE
Store at −20°C.Aliquot to avoid repeated freezing and thawing.
<0.005ng/μg (0.05EU/μg) as measured by LAL assay.
KITL
Activity has been measured by cell-based proliferation assay using TF-1 cells. The ED50 is typically < 12ng/mL.
Yes
Func, SDS-PAGE, WB-Re
Lyophilized from PBS, pH 7.2
20.6kDa
Chromatography
1mg
RUO
Clo, Con, Gb, Kitlg, Mgf, SCF, SF, SLF, Sl, Steel, contrasted
Kitl
Plants
His Tag
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit