Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse KLRG1 Alexa Fluor 700-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB6944N100UG
This item is not returnable.
View return policy
Description
KLRG1 Monoclonal specifically detects KLRG1 in Mouse samples. It is validated for Flow Cytometry.Specifications
KLRG1 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
2F1, CLEC15A, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFA, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600 | |
Chinese hamster ovary cell line CHO-derived mouse KLRG1 Glu57-Tyr188 Accession # O88713 | |
RUO | |
10219 | |
Mouse | |
IgG |
Flow Cytometry | |
1151A | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O88713 | |
Rabbit | |
100 μg | |
Primary | |
Detects mouse KLRG1 in flow cytometry. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction