Learn More
Abnova™ Mouse Lbp (Q61805) Full-length Recombinant Protein with His tag expressed in CHO cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4503
Catalog No. 89-950-437
Add to cart
Description
Description
Specifications
Specifications
Q61805
Lyophilized
16803
Lbp (Mouse) Recombinant Protein
10 ug
RUO
Ly88
Lbp
Hamster
His
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
64kDa
SDS-PAGE Stained with Coomassie Blue
Store at -20°C on dry atmosphere. After reconstitution with ultra-pure distilled H2O, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
<0.01ng/mL
Lbp
Up to 2μg/mL LBP mediates binding of FITC-LPS (0.5μg/mL) to CD14+CHO transfectants (2 x 106/mL).
Yes