Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Major Urinary Protein 3 Rat anti-Mouse, DyLight™ 680, Clone: SSI-7B10, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261648FR

Catalog No. NBP261648FR


Add to cart

Description

Description

Mouse Major Urinary Protein 3 Monoclonal antibody specifically detects Mouse Major Urinary Protein 3 in Mouse samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Mouse Major Urinary Protein 3
SSI-7B10
Western Blot, ELISA
Conjugated, Purified
Rat
IgG1 Kappa
100 μL
Primary
Mouse
ELISA, Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
MUP3
MUP3 purified from male NMRI mouse urine.
Protein A purified
Store at 4°C in the dark.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit