Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse MCP-1 (aa 24-96), Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

HEK293 cells Recombinant Protein

$250.50 - $770.50

Specifications

Recombinant Yes
Name Mouse MCP-1 (aa 24-96)
For Use With (Application) Control, ELISA, Western Blot
Concentration 200 µg/mL
Conjugate Unlabeled
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87900 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87900
10 µg
Each for $250.50
Add to cart
 
PIRP87901 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87901
25 µg
Each for $449.50
Add to cart
 
PIRP87902 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87902
100 µg
Each for $770.50
Add to cart
 
Description

Description

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Unlabeled
PBS with no preservative
HEK293 cells
-20°C
P10148
Ccl2
>95%
Mouse MCP-1 (aa 24-96)
200 µg/mL
Liquid
Acidic seminal fluid protein; AI323594; C-C motif chemokine 2; C-C motif chemokine ligand 2; Ccl2; chemokine (C-C motif) ligand 2; GDCF-2; HC11; H-MCP-1; HSMCR30; immediate-early serum-responsive JE protein; immediate-early serum-responsive protein JE; Je; MCAF; MCP1; MCP-1; MCP1A; MCP-1 A; MGC9434; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; monocyte chemotactic protein 1 A; Monocyte secretory protein JE; monocyte-chemoattractant protein-1 precursor; platelet-derived growth factor-inducible protein JE; RP23-350G1.3; Scya2; Sigje; small inducible cytokine A2; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; small inducible gene JE; small-inducible cytokine A2; SMC-CF
RUO
Mouse
20296
8.5 kD
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit