Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse MMP-9 ELISA Kit (Colorimetric)

Colorimetric ELISA Kit for detection of matrixmetalloprotinases

$611.50

Specifications

Species Mouse
Sensitivity 0.552ng/mL
Sample Type Serum, plasma, tissue homogenates
Research Discipline Cancer
Synonym 92kDa gelatinase, 92kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600951KT View Documents Novus Biologicals™
NBP2600951KIT
1 Kit Each for $611.50
Add to cart
 
Description

Description

The Mouse MMP-9 ELISA Kit (Colorimetric) measures mouse MMP-9 in serum, plasma, tissue homogenates.
Specifications

Specifications

Mouse
Serum, plasma, tissue homogenates
92kDa gelatinase, 92kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
MMP-9
4318
ELISA Kit (Colorimetric)
50 to 100μL
0.552ng/mL
Cancer
Store the unopened product at 2° to 8°C Protect from light. Do not use past expiration date.
ELISA
MMP9
0.625 to 40ng/mL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit