Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Monocyte Chemotactic Protein 5 ELISA Kit (Colorimetric)

Colorimetric ELISA Kit useful in study of Infection

$501.50

Specifications

Sensitivity 16.82pg/mL
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates
Research Discipline Immunology
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Target Monocyte Chemotactic Protein 5
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600961KT View Documents Novus Biologicals™
NBP2600961KIT
1 Kit Each for $501.50
Add to cart
 
Description

Description

The Mouse Monocyte Chemotactic Protein 5 ELISA Kit (Colorimetric) measures mouse Monocyte Chemotactic Protein 5 in serum, plasma, cell culture supernates,tissue homogenates.
Specifications

Specifications

16.82pg/mL
Immunology
Monocyte Chemotactic Protein 5
ELISA Kit (Colorimetric)
50 to 100 μL
Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
ELISA
15.6 to 1000pg/mL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit