Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse MOR31-2, Synthetic Peptide, Invitrogen™
Synthetic Peptide
Supplier: Thermo Scientific GSM00015P
Description
Mouse MOR31-2 Synthetic Peptide, GSM00015P, from Invitrogen.
GSM00015P is the immunizing peptide for the rabbit antibody to mouse olfactory receptor MOR31-2 (OSR00015W).GSM00015P may be used as a control in various immunology applications.Reconstitute with PBS or distilled water.Specifications
56860 | |
purified | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized | |
Control | |
Olfr690 | |
No preservative | |
50 μg |
Mouse | |
Mouse MOR31-2 | |
Synthetic Peptide | |
peptide with no preservative | |
ORL464, S18, Olfactory receptor Olfr690. GENE NAME: Olfr690 | |
Yes | |
Peptide | |
RUO - research use only |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title