Learn More
Raybiotech IncSupplier Diversity Partner Mouse Myeloperoxidase ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELMMPO1
For cell culture supernatants, plasma, and serum samples. Sensitivity: 600 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detect mouse MPO. Other species not determined yet
Catalog No. 50-195-0097