Learn More
Novus Biologicals™ Mouse Muscarinic Acetylcholine Receptor M2/CHRM2 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275277
Catalog No. NBP275277
Add to cart
Description
Description
The Mouse Muscarinic Acetylcholine Receptor M2/CHRM2 ELISA Kit (Colorimetric) measures mouse Muscarinic Acetylcholine Receptor M2/CHRM2 in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Mouse
1 Kit
This kit recognizes Mouse CHRM2 in samples. No significant cross-reactivity or interference between Mouse CHRM2 and analogues was observed.
1129
0.10 ng/mL
Muscarinic Acetylcholine Receptor M2/CHRM2
CHRM2
Biological Fluids, Plasma, Serum