Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Nicotinamide N-Methyltransferase/NNMT Recombinant Protein

Highly purified. Generating reliable and reproducible results.

$280.50 - $783.00

Specifications

Product Type Recombinant Protein
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term.
Gene Symbol Nnmt
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB59656100U View Documents Novus Biologicals™
NBP259656100UG
100μg Each for $783.00
Add to cart
 
NB5965620UG View Documents Novus Biologicals™
NBP25965620UG
20μg Each for $280.50
Add to cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-264 of Mouse Nicotinamide N-Methyltransferase/NNMT The Recombinant Mouse Nicotinamide N-Methyltransferase/NNMT Protein is derived from E. coli. The Recombinant Mouse Nicotinamide N-Methyltransferase/NNMT Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Recombinant Protein
RUO
Nnmt
Nnmt Recombinant Protein
In 20mM Tris-HCl (pH8.0) containing 10% glycerol,1mM DTT
E.coli
Unlabeled
Yes
Store at 4°C short term. Aliquot and store at −20°C long term.
FLJ21841, NES, Nestin
>90% by SDS-PAGE
Mouse
PAGE

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit