Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Osteoprotegerin/TNFRSF11B ELISA Kit (Colorimetric)

Colorimetric ELISA Kit useful in study of immunity, chemokine, chemotaxis

$602.00

Specifications

Sensitivity 3.9pg/mL
Sample Type Plasma, Serum, Tissue Homogenates
Research Discipline Cell Biology
Synonym OCIFMGC29565, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b
Storage Requirements Store unopened product at 2° to 8°C Protect from light. Do not use past expiration date.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601001KT View Documents Novus Biologicals™
NBP2601001KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Mouse Osteoprotegerin/TNFRSF11B ELISA Kit (Colorimetric) measures mouse Osteoprotegerin/TNFRSF11B in serum, plasma, tissue homogenates.
Specifications

Specifications

3.9pg/mL
Cell Biology
Store unopened product at 2° to 8°C Protect from light. Do not use past expiration date.
ELISA
TNFRSF11B
15.6 to 1000pg/mL
Plasma, Serum, Tissue Homogenates
OCIFMGC29565, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b
Osteoprotegerin/TNFRSF11B
4982
ELISA Kit (Colorimetric)
50 to 100 μL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit