Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Mouse PD-L1 / B7-H1 Protein;HEK293;100UG;PD1,PDL1,PD-L1,CD274,B7-H1,B7H1,B7-H,B7H,PDCD1L1,PDCD1LG1
Manufacturer:  ACROBiosystemsSupplier Diversity Partner PD1M5220100UG
Encompass_Preferred
Recombinant Protein;Mouse PD-L1 / B7-H1 Protein;100UG;Mouse PD-L1, His Tag (PD1-M5220) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Thr 238 (Accession # NP_068693).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.6 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse PD-L1, His Tag (Cat. No. PD1-M5220) at 10 μg/mL (100 μl/well) can bind Mouse PD-1, Fc Tag (Cat. No. PD1-M5259) with a linear range of 0.16-5 μg/mL (QC tested).
Catalog No. 50-108-9679