Learn More
Raybiotech IncSupplier Diversity Partner Mouse PDGF-AA ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELMPDGFAA1
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 3.2 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA pair antibody detects Mouse PDGF-AA and Human PDGF-AA.
Catalog No. 50-195-0107