Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse PLET-1 Alexa Fluor 700-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB6917N100UG
This item is not returnable.
View return policy
Description
PLET-1 Monoclonal specifically detects PLET-1 in Mouse samples. It is validated for Flow Cytometry.Specifications
PLET-1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8VEN2 | |
Plet1 | |
Mouse myeloma cell line NS0-derived recombinant mouse PLET-1 Ala27-Leu216 Accession # Q8VEN2 | |
RUO | |
349633 | |
Mouse | |
IgG1 |
Flow Cytometry | |
700738 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
AgK114, C11orf34, chromosome 11 open reading frame 34, placenta expressed transcript 1, PLET1 | |
Rat | |
100 μg | |
Primary | |
Detects mouse PLET-1 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction