Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Progesterone ELISA Kit (Colorimetric)

Colorimetric ELISA Kit useful in study of Cytokine

$602.00

Specifications

Sensitivity 0.2ng/mL
Sample Type Plasma, Serum
Research Discipline Signal Transduction
Synonym Pregn 4 ene 3 20 dione
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601251KT View Documents Novus Biologicals™
NBP2601251KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Mouse Progesterone ELISA Kit (Colorimetric) measures mouse Progesterone in serum, plasma.
Specifications

Specifications

0.2ng/mL
Signal Transduction
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
ELISA
0.3 to 10ng/mL
Plasma, Serum
Pregn 4 ene 3 20 dione
Progesterone
ELISA Kit (Colorimetric)
50 to 100 μL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit