Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse RAMP2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB6500
This item is not returnable.
View return policy
Description
RAMP2 Monoclonal specifically detects RAMP2 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
RAMP2 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
Q9WUP0 | |
RAMP2 | |
E. coli-derived recombinant mouse RAMP2 Ser45-Val159 Accession # Q9WUP0 | |
100 μg | |
Primary | |
Detects mouse RAMP2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human RAMP2 is observed. | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
773816 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
calcitonin receptor-like receptor activity modifying protein 2, Calcitonin-receptor-like receptor activity-modifying protein 2, CRLR activity-modifying protein 2, receptor (calcitonin) activity modifying protein 2, receptor (G protein-coupled) activity modifying protein 2, receptor activity modifying protein 2, receptor activity-modifying protein 2, receptor-activity-modifying protein 2 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10266 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction