Learn More
Novus Biologicals™ Mouse SAA2 ELISA Kit (Colorimetric)
Mouse SAA2 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276738
Catalog No. NBP276738
Add to cart
Description
Description
The Mouse SAA2 ELISA Kit (Colorimetric) measures mouse SAA2 in serum, plasma and other biological fluids.
Specifications
Specifications
31.25 to 2000 pg/mL
ELISA Kit (Colorimetric)
Storage is content dependent.
SAA2
Biological Fluids, Plasma, Serum
6289
18.75 pg/mL
Mouse
1 Kit