Learn More
Raybiotech IncSupplier Diversity Partner Mouse sFRP-3 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELMSFRP31
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 25 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse sFRP-3.
Catalog No. 50-195-0131