Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Novus Biologicals™ Mouse STAT5A ELISA Kit (Colorimetric)
This kit recognizes Mouse STAT5A in samples.
Manufacturer:  Novus Biologicals™ NBP280281
Catalog No. NB013352
Description
Description
No significant cross-reactivity or interference between Mouse STAT5A and analogues was observed.
Specifications
Specifications
P42230, P42230
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent
Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
1 Kit
Serum, plasma, and other biological fluids
31.25 to 2000 pg/mL
6776
18.75 pg/mL
Mouse
STAT5A
STAT5A