Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Stella/Dppa3 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | Stella/Dppa3 |
---|---|
Clone | 283910 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Description
Stella/Dppa3 Monoclonal specifically detects Stella/Dppa3 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Stella/Dppa3 | |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Rat | |
Mouse | |
Q8QZY3 | |
359787 | |
E. coli-derived recombinant mouse Stella/Dppa3 Met1-Asn150 Accession # Q8QZY3 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Stella/Dppa3 in direct ELISAs and Western blots. |
283910 | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
developmental pluripotency associated 3, DPPA3, DPPA3 developmental pluripotency associated 3 | |
DPPA3 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title