Learn More
Novus Biologicals™ Mouse Surfactant Protein A ELISA Kit (Colorimetric)
Mouse Surfactant Protein A ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276693
Catalog No. NBP276693
Add to cart
Description
Description
The Mouse Surfactant Protein A ELISA Kit (Colorimetric) measures mouse Surfactant Protein A in serum, plasma and other biological fluids.
Specifications
Specifications
0.313 to 20 ng/mL
ELISA Kit (Colorimetric)
Storage is content dependent.
SFTPA1
Biological Fluids, Plasma, Serum
653509
0.188 ng/mL
Mouse
1 Kit