Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Tau ELISA Kit (Colorimetric)

This kit recognizes Mouse Tau in samples.

$629.00

Specifications

Assay Range 15.63 to 1000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 4137
Gene Symbol MAPT
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Interassay CV Intraassay CV Price Quantity & Availability  
Catalog Number Mfr. No. Interassay CV Intraassay CV Price Quantity & Availability  
NB016722
View Documents
Novus Biologicals™
NBP281163
0.0472 0.0551
Each for $629.00
Add to cart
 
NB016819
View Documents
Novus Biologicals™
NBP281260
0.0527 0.0466
Each for $629.00
Add to cart
 
Description

Description

No significant cross-reactivity or interference between Mouse Tau and analogues was observed.
Specifications

Specifications

15.63 to 1000 pg/mL
4137
Mouse
1 Kit
Serum, Plasma, Cell Supernatant, Other Biological Fluids
Storage is content dependent
ELISA
MAPT
ELISA Kit (Colorimetric)
Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Markers
9.38 pg/mL
Tau
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit