Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse Thrombin-antithrombin (TAT) Complexes ELISA Kit (Colorimetric)

Highly purified and high bioactivity. Generating reliable and reproducible results.

Manufacturer:  Novus Biologicals™ NBP275962

Catalog No. NBP275962


Add to cart

Description

Description

  • The Mouse Thrombin-antithrombin (TAT) complexes ELISA Kit (Colorimetric) measures mouse Thrombin-antithrombin (TAT) complexes in plasma, cell culture.
  • This assay employs a quantitative enzyme immunoassay technique that measures the specified antigen in samples.
  • Standard Curve Range: 0.031-8 ng/mL (lot dependent)
  • Sensitivity: 0.01 ng/mL (lot dependent)
  • Assay Type: Sandwich ELISA
  • Inter-Assay: 9.90% (lot dependent)
  • Intra-Assay: 5.00% (lot dependent)
  • Spike Recovery: 97.00% (lot dependent)
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Specifications

Specifications

ELISA
1 Kit
Storage is content dependent
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit