Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Thrombopoietin (aa 22-356), Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

CHO cells Recombinant Protein

$238.00 - $1092.00

Specifications

Recombinant Yes
Name Mouse Thrombopoietin (aa 22-356)
For Use With (Application) Control, ELISA, Western Blot
Concentration 200 µg/mL
Conjugate Unlabeled
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87909 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87909
10 µg
Each for $238.00
Add to cart
 
PIRP87910 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87910
25 µg
Each for $417.50
Add to cart
 
PIRP87911 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87911
100 µg
Each for $1,092.00
Add to cart
 
Description

Description

Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, ultimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Unlabeled
PBS with no preservative; pH 7.2
CHO cells
-20°C
P40226
THPO
>95%
Mouse Thrombopoietin (aa 22-356)
200 µg/mL
Liquid
C-mpl ligand; megakaryocyte colony-stimulating factor; Megakaryocyte growth and development factor; megakaryocyte stimulating factor; MGDF; MKCSF; ML; MPL ligand; Mpllg; Myeloproliferative leukemia virus oncogene ligand; prepro-thrombopoietin; THCYT1; THPO; thrombopoietin; TPO
RUO
Mouse
21832
35.5 kD
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit