Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Mouse Thyroxine ELISA Kit (Colorimetric)
Colorimetric ELISA Kit useful in study of thyroid functions
Supplier: Novus Biologicals™ NBP2601621KIT
Description
The Mouse Thyroxine ELISA Kit (Colorimetric) measures mouse Thyroxine in serum, plasma, tissue homogenates.Specifications
20 to 320ng/mL | |
ELISA Kit (Colorimetric) | |
Detection of mouse T4 | |
ELISA | |
1 Kit | |
Signal Transduction | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. | |
Thyroxine |
20ng/mL | |
Plasma, Serum, Tissue Homogenates | |
Mouse | |
Assay plate, Standard, Conjugate, HRP-avidin, Wash Buffer (20X concentrate), Substrate A, Substrate B, Stop Solution, Adhesive Strip (For 96 wells), Instruction manual | |
RUO | |
50 to 100 μL | |
T4 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction