Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Mouse TNF-alpha Quantikine ELISA Kit
Mouse TNF-alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
$4199.20
Specifications
Assay Range | 10.90 - 700pg/mL |
---|---|
Assay Sensitivity | 7.21pg/mL |
Conjugate | HRP |
Product Type | Solid Phase Sandwich ELISA |
Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum |
Catalog Number | Mfr. No. | Quantity | Sufficient For | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Sufficient For | Price | Quantity & Availability | ||||
SMTA00B
|
R&D Systems™
SMTA00B |
6 Plates | Sufficient materials to run ELISAs on at least six 96-well plates |
Each for $4,199.20
|
|
||||
MTA00B
|
R&D Systems™
MTA00B |
2 Plates | Sufficient materials to run ELISAs on at least two 96-well plates | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
The Mouse TNF-alpha Quantikine ELISA Kit quantifies mouse TNF-alpha in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).Specifications
10.90 - 700pg/mL | |
HRP | |
Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum | |
Mouse | |
21926 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
7.21pg/mL | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Mouse TNF-alpha | |
ELISA | |
Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL) | |
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title