Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse TNF RI/TNFRSF1A ELISA Kit (Colorimetric)

Colorimetric ELISA Kit is useful in study of Thyroid function

$602.00

Specifications

Sensitivity 1.261pg/mL
Sample Type Plasma, Serum, Tissue Homogenates
Research Discipline Signal Transduction
Synonym CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601671KT View Documents Novus Biologicals™
NBP2601671KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Mouse TNF RI/TNFRSF1A ELISA Kit (Colorimetric) measures mouse TNF RI/TNFRSF1A in serum, plasma, tissue homogenates.
Specifications

Specifications

1.261pg/mL
Signal Transduction
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
ELISA
TNFRSF1A
15.625 to 1000pg/mL
Plasma, Serum, Tissue Homogenates
CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor
TNF RI/TNFRSF1A
7132
ELISA Kit (Colorimetric)
50 to 100 μL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit