Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse TNF RII/TNFRSF1B ELISA Kit (Colorimetric)

Assay has high sensitivity and specificity for detection of mouse TNFsR- II

$602.00

Specifications

Species Mouse
Sensitivity 1.6pg/mL.
Sample Type serum, plasma, tissue homogenates
Research Discipline Signal Transduction
Specificity For detection of mouse TNFsR- II
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601291KT View Documents Novus Biologicals™
NBP2601291KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Mouse TNF RII/TNFRSF1B ELISA Kit (Colorimetric) measures mouse TNF RII/TNFRSF1B in serum, plasma, tissue homogenates.
Specifications

Specifications

Mouse
serum, plasma, tissue homogenates
For detection of mouse TNFsR- II
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
ELISA
TNFRSF1B
7.8125 pg/ml to 500 pg/ml
1.6pg/mL.
Signal Transduction
CD120b, CD120b antigen, Etanercept, p75 TNF receptor, p75TBPII, p75TNFR, soluble TNFR1B variant 1, TNFBRp80 TNF-alpha receptor, TNF-R2, TNFR2TNFR1B, TNF-R75, TNFR80, TNF-RII, TNF-R-II, TNFR-II, tumor necrosis factor beta receptor, tumor necrosis factor binding protein 2, Tumor necrosis factor receptor 2, tumor necrosis factor receptor superfamily member 1B, tumor necrosis factor receptor superfamily, member 1B, Tumor necrosis factor receptor type II
TNF RII/TNFRSF1B
7133
ELISA Kit (Colorimetric)
Adhesive Strip (For 96 wells), Assay plate (12 x 8 coated Microwells), Biotin-antibody (100 x concentrate), Biotin-antibody Diluent, HRP-avidin Diluent, HRP-avidin (100 x concentrate), TMB Substrate, Wash Buffer (25 x concentrate), Stop Solution, Standard (Freeze dried), Sample Diluent, Instruction manual

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit