Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Mouse Toxoplasmosis (TOXO) Antibody (IgG) ELISA Kit (Colorimetric)
Assay has high sensitivity and specificity for detection of mouse anti-toxoplasmosis (TOXO) antibody (IgG).
Supplier: Novus Biologicals™ NBP2601691KIT
Description
The Mouse Toxoplasmosis (TOXO) Antibody (IgG) ELISA Kit (Colorimetric) measures mouse Toxoplasmosis (TOXO) Antibody (IgG) in serum.Specifications
Qualitative | |
Serum | |
Mouse | |
Negative Control, Positive Control, Instruction manual, Sample Diluent, Stop Solution, Substrate B, Substrate A, Wash Buffer (30 x concentrate), Coated assay plate, HRP-conjugate, Adhesive Strip (For 96 wells) | |
Cell Biology | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. |
ELISA Kit (Colorimetric) | |
For detection of mouse anti-toxoplasmosis (TOXO) antibody (IgG) | |
ELISA | |
1 Kit | |
50 to 100 μL | |
Toxoplasmosis (TOXO) Antibody (IgG) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction