Learn More
Crystal Chem IncSupplier Diversity Partner Mouse Total Bile Acids Assay Kit, 5-Kit Pack, 96-test per kit DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80475
The Mouse Total Bile Acidsl Assay is used to quantify levels of total bile acids in mouse serum and plasma in less than 10 minutes. The 5-Kit Pack is convieniently provided as five individual kits.Highlights: Kit uses sample size of only 20 µL. Excellent correlation to ELISA method. Fast, 10 minute procedureProtocol for liver and feces samples available. Designed for mouse samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7716