Learn More
Raybiotech IncSupplier Diversity Partner Mouse VEGFR2 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELMVEGFR21
For cell culture supernatants, plasma, and serum samples. Sensitivity: 80 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30L, CD30, CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN-gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 R beta, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN (OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, TARC, TCA-3, TECK, TIMP-1, TNF RI, TNF RII, TPO, VCAM-1.
Catalog No. 50-195-0163