Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOV10L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551004
Description
MOV10L1 Polyclonal Antibody detects endogenous levels of MOV10L1 protein.
MOV10L1 Polyclonal antibody specifically detects MOV10L1 in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
MOV10L1 | |
Polyclonal | |
Unconjugated | |
MOV10L1 | |
Cardiac helicase activated by MEF2 protein; cardiac helicase activated by MEF2C protein; cardiac-specific RNA helicase; Champ; Csm; DJ402G11.8; Moloney leukemia virus 10-like 1; moloney leukemia virus 10-like protein 1; Moloney leukemia virus 10-like protein 1 homolog; Mov10 RISC complex RNA helicase like 1; Mov10l1; Mov10l1, Moloney leukemia virus 10-like 1, homolog; Mov10-like 1; MOV10-like protein 1; MOV10-like protein 1 homolog; putative helicase Mov10l1; RNA helicase Mov10l1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54456 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9BXT6 | |
MOV10L1 | |
Synthesized peptide derived from the Internal region of human MOV10L1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction