Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moxalactam sodium salt, 50 mg/mL in distilled water, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
5 x 1mL
64953-12-4
C20H18N6Na2O9S
564.437
MFCD03427565
GRIXGZQULWMCLU-GDUWRUPCSA-L
12856838
disodium;(6R,7R)-7-[[(2R)-2-carboxylato-2-(4-hydroxyphenyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-oxa-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C(C4=CC=C(C=C4)O)C(=O)[O-])OC)OC2)C(=O)[O-].[Na+].[Na+]
64953-12-4
C20H18N6Na2O9S
564.437
MFCD03427565
GRIXGZQULWMCLU-GDUWRUPCSA-L
12856838
disodium;(6R,7R)-7-[[(2R)-2-carboxylato-2-(4-hydroxyphenyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-oxa-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)C(C4=CC=C(C=C4)O)C(=O)[O-])OC)OC2)C(=O)[O-].[Na+].[Na+]
Physical Form Liquid
Concentration 50mg/mL in distilled water
Quantity 5 x 1mL
Chemical Name or Material Moxalactam sodium salt, sterile-filtered

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

storageNote1 : −30° to −10°C