Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Moxifloxacin hydrochloride, 97%

Click to view available options
Quantity:
100mg
500mg
186826-86-8
C21H25ClFN3O4
437.896
MFCD00949117
IDIIJJHBXUESQI-DFIJPDEKSA-N
101526
CHEBI:7008
7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid;hydrochloride
COC1=C2C(=CC(=C1N3CC4CCCNC4C3)F)C(=O)C(=CN2C5CC5)C(=O)O.Cl
186826-86-8
C21H25ClFN3O4
437.896
MFCD00949117
IDIIJJHBXUESQI-DFIJPDEKSA-N
101526
CHEBI:7008
7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carboxylic acid;hydrochloride
COC1=C2C(=CC(=C1N3CC4CCCNC4C3)F)C(=O)C(=CN2C5CC5)C(=O)O.Cl
Applications
Moxifloxacin hydrochloride is a broad spectrum antibiotic commonly used against bacteria which cause sinus and lung infections. Mitoxantrone is a cytostatic anthracenedione that intercalates in DNA and increases the incidence of double-strand breaks by stabilizing the cleavable complex of topoisomerase II and DNA. Mitoxantrone also displays broad immunosuppressive activity inhibiting proliferation of all classes of lymphocytes and inducing apoptosis of antigen-presenting T cells. It used clinically as a chemotherapeutic agent against leukemias and solid tumors and as an immune system modulator in multiple sclerosis.

Solubility
Sparingly soluble in water. Soluble in DMSO

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Beilstein 8377447
Odor Odorless
Solubility Information Sparingly soluble in water; Soluble in DMSO
Formula Weight 437.89
Physical Form Powder
Percent Purity 97%
Boiling Point 636°C
Color Yellow
Quantity 100mg
Chemical Name or Material Moxifloxacin hydrochloride

RTECSNumber : VB1983750

storageNote1 : 2° to 8°C