Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOZ, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548342

Catalog No. PA548342

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Histone acetyltransferase that acetylates lysine residues in histone H3 and histone H4 (in vitro). Component of the MOZ/MORF complex which has a histone H3 acetyltransferase activity. May act as a transcriptional coactivator for RUNX1 and RUNX2. Acetylates p53/TP53 at 'Lys-120' and 'Lys-382' and controls its transcriptional activity via association with PML.

Specifications

Specifications

MOZ
Polyclonal
PBS with 0.09% sodium azide
Q8BZ21
Histone acetyltransferase KAT6A, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 3, MYST-3, Monocytic leukemia zinc finger protein, Runt-related transcription factor-binding protein 2, Zinc finger protein 220, KAT6A, MOZ, MYST3, RUNXBP2, ZNF220
Rabbit
IgG
400μL
Store at 2° to 8°C for up to 6 months. For long term storage store at -20°C.
Primary
244349
Western Blot
Unconjugated
Kat6a
Liquid
KAT6A
KLH conjugated synthetic peptide between 1319-1351 amino acids from the C-terminal region of human MOZ/MYST3.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit