Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin 35% Solution

Globulin free albumins are suitable for use in applications where no other proteins should be present

$387.00 - $2100.00

Specifications

Concentration 34.4g/dL
Format Solution
For Use With (Application) Immunoassays, Mammalian Cell Culture
Molecular Weight ∽66
Name Bovine Albumin 35% Solution
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN810061
View Documents
MP Biomedicals™
08810061
50mL
Each for $387.00
Add to cart
 
ICN810063
View Documents
MP Biomedicals™
08810063
1L
Each for $2,100.00
Add to cart
 
Description

Description

Albumins are a group of acidic proteins which occur plentifully in the body fluids and tissues of mammals and in some plant seeds.

  • CAS Number: 9048-46-8
  • MDL Number: MFCD00130384
  • Format: Solution
  • Sterile albumins
  • Concentration: About 35g/dl

For immunoassays and mammalian cell culture. Used to solubilize lipids and is also used as a blocking agent in Western blots or ELISA applications.

Specifications

Specifications

34.4g/dL
Immunoassays, Mammalian Cell Culture
Bovine Albumin 35% Solution
0° to 5°C
Solution
∽66
7.2
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit