Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Amylase
Derived from Aspergillus oryzae
Supplier: MP Biomedicals™ 0215037501
Description
- Format: Tan powder
- Specific Activity: 50 Units/mg protein
Specifications
>50 U/mg | |
4.8 to 5.5 | |
Aspergillus oryzae | |
alpha-Amylase |
Tan | |
0°C | |
1 mU | |
Powder |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction