Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ (Arg8)-Vasopressin
$120.00 - $474.00
Specifications
For Use With (Application) | Cell Biology and Analysis |
---|---|
Molecular Weight (g/mol) | 1084.23 |
Name | (Arg-8)-Vasopressin (AVP) |
Storage Requirements | -20°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ICN15296201
|
MP Biomedicals™
0215296201 |
1mg |
Each for $120.00
|
|
|||||
ICN15296205
|
MP Biomedicals™
0215296205 |
5mg |
Each for $474.00
|
|
|||||
Description
- Contains a disulfide bond between Cys1 and Cys6
Specifications
Cell Biology and Analysis | |
1084.23 | |
(Arg-8)-Vasopressin (AVP) | |
-20°C |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title