Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ BCIP/NBT Liq. Substrate, 100mL
Stabilized Chromogen Solution specially prepared for immunoblotting procedures only
Supplier: MP Biomedicals™ 08980871
Specifications
BCIP or NBT | |
Immunoblotting procedures | |
0° to 5°C |
Liquid | |
BCIP/NBT Substrate | |
100mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction