Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals beta-Glucosidase from Sweet Almonds
Involved in hydrolysis of β-glycosidic bonds connecting carbohydrate residues in β-D-glycosides
Manufacturer: MP Biomedicals 0210034894
Description
- Synonyms: β-D-Glucoside glucohydrolase
- CAS Number: 9001-22-3
- EC Number: 232-589-7
- MDL Number: MFCD00130628
- Format: Lyophilized powder, salt free
- Specific Activity: >2500U/mg solid
- Dissolves readily at 5mg/mL in 0.1M sodium acetate (pH 5.0) to give clear pale yellow solution
- Involved in synthesis of glucosides and fucosides
- Used in pharmaceutical, cosmetic and detergent industries
- Hydrolytic removal of aglycone moiety from flavonoid and isoflavonoid glycosides
- Flavor enhancement of fruit juices and wine
- Biosynthesis of oligosaccharides
Specifications
beta-Glucosidase | |
>2,500U/mg | |
Sweet almonds | |
Potential applications in pharmaceutical, cosmetic and detergent industries, hydrolytic removal of aglycone moiety from Flavonoid and Isoflavonoid Glycosides, flavor enhancement of fruit juices and wine and biosynthesis of Oligosaccharides |
Lyophilized Powder | |
Pale Yellow | |
5000kU | |
-20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title