Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ beta-Glucosidase from Sweet Almonds

Involved in hydrolysis of β-glycosidic bonds connecting carbohydrate residues in β-D-glycosides

Specifications

Color Pale Yellow
Concentration >2500 U/mg
Product Type beta-Glucosidase
Format Lyophilized Powder
For Use With (Application) Potential applications in pharmaceutical, cosmetic and detergent industries, hydrolytic removal of aglycone moiety from Flavonoid and Isoflavonoid Glycosides, flavor enhancement of fruit juices and wine and biosynthesis of Oligosaccharides
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN10034890
View Documents Promotion Details
MP Biomedicals™
0210034890
500 kU This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Synonyms: β-D-Glucoside glucohydrolase
  • CAS Number: 9001-22-3
  • EC Number: 232-589-7
  • MDL Number: MFCD00130628
  • Format: Lyophilized powder, salt free
  • Specific Activity: >2500U/mg solid
  • Dissolves readily at 5mg/mL in 0.1M sodium acetate (pH 5.0) to give clear pale yellow solution

  • Involved in synthesis of glucosides and fucosides
  • Used in pharmaceutical, cosmetic and detergent industries
  • Hydrolytic removal of aglycone moiety from flavonoid and isoflavonoid glycosides
  • Flavor enhancement of fruit juices and wine
  • Biosynthesis of oligosaccharides

Specifications

Specifications

Pale Yellow
beta-Glucosidase
Potential applications in pharmaceutical, cosmetic and detergent industries, hydrolytic removal of aglycone moiety from Flavonoid and Isoflavonoid Glycosides, flavor enhancement of fruit juices and wine and biosynthesis of Oligosaccharides
>2500 U/mg
Lyophilized Powder
Sweet almonds
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit