Learn More
MP Biomedicals™ Mycoplasma Hoechst Stain Kit
Highly sensitive test detects mycoplasma and other prokaryotic organisms in under two hours
Manufacturer:  MP Biomedicals™ 093030000
Catalog No. ICN3030000
Add to cart
Description
Description
  • Uses DNA fluorochrome staining technology with Hoechst Stain 33258
  • For the in situ detection of mycoplasma, bacteria, yeast, and fungi in cell cultures
  • Testing is rapid (less than 2 hours)and economical (100 tests per kit)
  • Non-nuclear staining is readily apparent, with contaminants standing out boldly against a black background
Specifications
Specifications
15° to 30°C
100 Tests
Mycoplasma Stain Kit